Gene Transcripts
- Gene Symbol
- Derl3
- Entrez Gene ID
- 70377
- Gene Name
- Der1-like domain family, member 3
- Gene Aliases
- 1810006I20Rik, 1810063P04Rik, IZP6, derlin-3
- Chromosome Location
- Chr. 10: 75893080 - 75895941 on Build GRCm38
- UniGene ID
- Mm.275878
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024440.2 |
NP_077760.1
|
5, 6 |
560 |
|
GenBank mRNA |
AB047555.1 |
- |
5, 6 |
560 |
|
AK007348.1 |
- |
5, 6 |
422 |
|
AK007948.1 |
- |
5, 6 |
546 |
|
AK172429.1 |
- |
5, 6 |
546 |
|
BC004729.1 |
- |
5, 6 |
543 |
|
BC019815.1 |
- |
5, 6 |
540 |
|
BC099615.1 |
- |
5, 6 |
540 |
|
BC115466.1 |
- |
5, 6 |
555 |
40100387 |
BC115467.1 |
- |
5, 6 |
555 |
40100388 |
View Full Product Details