Gene Transcripts

Gene Symbol
Satl1
Entrez Gene ID
73809
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases
4930404K22Rik
Chromosome Location
Chr. X: 112383207 - 112406779 on Build GRCm38
UniGene ID
Mm.50508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028655.1 NP_082931.1 1 1378
GenBank mRNA AK015086.1 - 1 421
BC138009.1 - 1 1378 8861028
BC138011.1 - 1 1378 8861030

View Full Product Details