Gene Transcripts

Gene Symbol
Syce3
Entrez Gene ID
75459
Gene Name
synaptonemal complex central element protein 3
Gene Aliases
1700007E06Rik, TSEG-2, TSEG2
Chromosome Location
Chr. 15: 89390174 - 89410503 on Build GRCm38
UniGene ID
Mm.45462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162880.1 NP_001156352.1 2 170
NM_001162882.1 NP_001156354.1 2 138
GenBank mRNA AK005702.1 - 2 92
BC048615.1 - 2 170
EU079025.1 - 2 131
HQ130280.1 - 1 65

View Full Product Details