Gene Transcripts
- Gene Symbol
- Tmem50b
- Entrez Gene ID
- 77975
- Gene Name
- transmembrane protein 50B
- Gene Aliases
- AU015466, AU019872, B230114J08Rik
- Chromosome Location
- Chr. 16: 91574508 - 91597680 on Build GRCm38
- UniGene ID
- Mm.290341
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030018.3 |
NP_084294.1
|
4, 5 |
427 |
|
GenBank mRNA |
AK020977.1 |
- |
4, 5 |
432 |
|
AK098154.1 |
- |
4, 5 |
391 |
|
AK133966.1 |
- |
4, 5 |
455 |
|
AK150773.1 |
- |
|
415 |
|
AK151796.1 |
- |
4, 5 |
457 |
|
AK152190.1 |
- |
4, 5 |
443 |
|
AK152767.1 |
- |
4, 5 |
443 |
|
AK155082.1 |
- |
4, 5 |
480 |
|
AK170684.1 |
- |
4, 5 |
422 |
|
AK172335.1 |
- |
4, 5 |
415 |
|
BC004841.1 |
- |
4, 5 |
430 |
2649264 |
View Full Product Details