Product Details

SNP ID
rs13478559
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:140442021 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTGTGGAAGGCTTGCCCAGTG[C/T]ACAAGCACTGACATCTTCCAGACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Eif3b PubMed Links

Gene Details

Gene
Eif3b
Gene Name
eukaryotic translation initiation factor 3, subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133916.2 Intron NP_598677.1

View Full Product Details