Product Details

SNP ID
rs13478767
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:55224284 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGGACAGCCCACTGGGGTACA[A/G]CCACACAGAGGGGAATTCCAGGATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Fam188b PubMed Links

Gene Details

Gene
Fam188b
Gene Name
family with sequence similarity 188, member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142781.1 1077 Missense Mutation NP_001136253.1
NM_177883.4 1077 Missense Mutation NP_808551.2

View Full Product Details