Product Details
- SNP ID
-
rs13481258
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118479301 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGAGGACAGGTGAGGCTCTGGT[A/G]CAGTGCTGGTCTTATTACTGGCCAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Engase
PubMed Links
Gene Details
- Gene
- Engase
- Gene Name
- endo-beta-N-acetylglucosaminidase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172573.2 |
|
Intron |
|
|
NP_766161.1 |
View Full Product Details