Product Details
- SNP ID
-
rs6191597
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:62583867 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGACACATCAGGACTTAGTGTGAC[C/T]TGTCTGCCAAAGAAATGACTTTGGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gpr149
PubMed Links
Gene Details
- Gene
- Gpr149
- Gene Name
- G protein-coupled receptor 149
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177346.4 |
|
Intron |
|
|
NP_796320.2 |
View Full Product Details