Product Details

SNP ID
rs13478097
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:5682750 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGTGTAATGTTCTCTATGGAT[A/G]GCCACGAATATTATATTTGTTTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Steap2 PubMed Links

Gene Details

Gene
Steap2
Gene Name
six transmembrane epithelial antigen of prostate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103156.2 621 Silent Mutation NP_001096626.1
NM_001103157.2 621 Silent Mutation NP_001096627.1
NM_001285469.1 621 Silent Mutation NP_001272398.1
NM_001285470.1 621 Silent Mutation NP_001272399.1
NM_001285471.1 621 Silent Mutation NP_001272400.1
NM_028734.5 621 Silent Mutation NP_083010.2

View Full Product Details