Product Details
- SNP ID
-
rs13476464
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:43682830 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCCGAGTGTTGCACACATATCT[A/T]TGTTAACATAGCAAAGCTTTCCACT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Kynu
PubMed Links
Gene Details
- Gene
- Kynu
- Gene Name
- kynureninase (L-kynurenine hydrolase)
There are no transcripts associated with this gene.
View Full Product Details