Product Details
- SNP ID
-
rs3690107
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:145879002 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATATACCCCTCCATGTGTCAGAG[G/A]AACTGGACAGCGAGGTTTTGGGGGA
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gm39954
PubMed Links
Gene Details
- Gene
- Gm39954
- Gene Name
- predicted gene, 39954
There are no transcripts associated with this gene.
- Gene
- Rin2
- Gene Name
- Ras and Rab interactor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028724.4 |
|
Intron |
|
|
NP_083000.5 |
View Full Product Details