Product Details
- SNP ID
-
rs13478715
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:37112944 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTCTTCAACACAAGCTTTAGGAG[C/G]AAATGCTGGACCCAAACCACAATAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Dgki
PubMed Links
Gene Details
- Gene
- Dgki
- Gene Name
- diacylglycerol kinase, iota
View Full Product Details