Product Details

SNP ID
rs13483042
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:55498625 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTGTGACTGGGCAATTATTCCACCC[C/T]ACAATGCACTGATGGGCTGTCTATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
St6gal2 PubMed Links

Gene Details

Gene
St6gal2
Gene Name
beta galactoside alpha 2,6 sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172829.2 1677 UTR 3 NP_766417.1

View Full Product Details