Product Details
- SNP ID
-
rs13479746
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:48603491 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACAGAGTCATTGTCACCAGACC[A/G]TCCTTGCTCAGAGTTCTACTTTCAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tenm3
PubMed Links
Gene Details
- Gene
- Tenm3
- Gene Name
- teneurin transmembrane protein 3
View Full Product Details