Product Details
- SNP ID
-
rs6407687
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:105266062 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTACTTGGTAGACAAGTAGTTT[G/T]TTGACTAAGCCAGGCCCTAGGGTGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Tlk2
PubMed Links
Gene Details
- Gene
- Tlk2
- Gene Name
- tousled-like kinase 2 (Arabidopsis)
There are no transcripts associated with this gene.
View Full Product Details