Product Details

SNP ID
rs13480599
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:43906118 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACCTTCTTGCAAGTTTACAACTAA[A/G]TAGTGACAAGGCTGAGTGTCCACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Qrsl1 PubMed Links

Gene Details

Gene
Qrsl1
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
There are no transcripts associated with this gene.

Gene
Rtn4ip1
Gene Name
reticulon 4 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130892.4 Intron NP_570962.2

View Full Product Details