Product Details

SNP ID
rs4173891
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:37212868 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGGGAGGTGGCCAGCAGGGAT[A/T]TGGTAGGAGGGTTCTGTAGGCAGAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Stxbp5l PubMed Links

Gene Details

Gene
Stxbp5l
Gene Name
syntaxin binding protein 5-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114611.1 Intron NP_001108083.1
NM_001114612.1 Intron NP_001108084.1
NM_001114613.1 Intron NP_001108085.1
NM_172440.3 Intron NP_766028.2

View Full Product Details