Product Details
- SNP ID
-
rs13476942
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:180687637 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCTGCACCCAGAGTCCTGCTCAT[C/T]GGTGGCGCTACCGTTTGTCTCATCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dido1
PubMed Links
Gene Details
- Gene
- Dido1
- Gene Name
- death inducer-obliterator 1
There are no transcripts associated with this gene.
View Full Product Details