Product Details

SNP ID
rs13476571
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:72401801 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAATCGGGCTCAGTGTGGTGCTTTG[C/G]GAGATCTCAGTTATGATTTACCCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Zak PubMed Links

Gene Details

Gene
Zak
Gene Name
sterile alpha motif and leucine zipper containing kinase AZK
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164791.1 Intron NP_001158263.1
NM_023057.5 Intron NP_075544.1
NM_178084.4 Intron NP_835185.2

View Full Product Details