Product Details
- SNP ID
-
rs4223226
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74719881 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACTCTCTGCCAGATCCCTATTCT[C/T]TCTCTTTCATTCACAAAACCTGGGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Hoxd3
PubMed Links
Gene Details
- Gene
- Hoxd3
- Gene Name
- homeobox D3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010468.2 |
|
Intron |
|
|
NP_034598.2 |
- Gene
- Hoxd3os1
- Gene Name
- homeobox D3, opposite strand 1
There are no transcripts associated with this gene.
- Gene
- Hoxd4
- Gene Name
- homeobox D4
There are no transcripts associated with this gene.
- Gene
- Mir10b
- Gene Name
- microRNA 10b
There are no transcripts associated with this gene.
View Full Product Details