Product Details

SNP ID
rs13481122
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:81625525 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTTAGCATCCTCTCAGGTTTC[A/T]TCCTCTGGACAAAGCAGATTCCCTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Asic2 PubMed Links

Gene Details

Gene
Asic2
Gene Name
acid-sensing (proton-gated) ion channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034013.2 Intron NP_001029185.1
NM_007384.3 Intron NP_031410.1

View Full Product Details