Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDC3
Entrez Gene ID
101926635
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
YJDC
Chromosome Location
Chr.7: 52670941 - 52734016 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285245.1 NP_001272174.1 6 - 7 1270 77
XM_005560075.2 XP_005560132.1 6 - 7 1752 77
XM_005560076.2 XP_005560133.1 7 - 8 1490 77
XM_015452452.1 XP_015307938.1 7 - 8 1506 77
XM_015452453.1 XP_015307939.1 7 - 8 1431 77
GenBank mRNA AB168338.1 - 6 - 7 1270 77

View Full Product Details