Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIPPLY1
Entrez Gene ID
102146788
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.X: 104022697 - 104029030 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594295.2 XP_005594352.1 2 - 3 3355 82

View Full Product Details