Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNCN
Entrez Gene ID
102131315
Gene Name
kinocilin
Gene Aliases
-
Chromosome Location
Chr.1: 180951214 - 180955365 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543479.2 XP_005543536.1 2 - 3 388 61

View Full Product Details