Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APLN
Entrez Gene ID
102144427
Gene Name
apelin
Gene Aliases
-
Chromosome Location
Chr.X: 126520388 - 126530850 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594539.2 XP_005594596.1 1 - 2 1173 61

View Full Product Details