Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPP7
Entrez Gene ID
102143702
Gene Name
phospholipid phosphatase 7 (inactive)
Gene Aliases
PPAPDC3
Chromosome Location
Chr.15: 7647446 - 7663959 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580612.2 XP_005580669.1 2 - 3 897 92

View Full Product Details