Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC12
Entrez Gene ID
102127860
Gene Name
sialic acid binding Ig like lectin 12
Gene Aliases
-
Chromosome Location
Chr.19: 52232563 - 52243163 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590115.2 XP_005590172.1 1 - 2 497 97

View Full Product Details