The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007464.3 | NP_031490.2 | 1 - 2 | 350 | 61 |
GenBank mRNA | AK156097.1 | - | 1 - 2 | 39 | 61 |
AK172636.1 | - | 1 - 2 | 83 | 61 | |
BC011338.1 | - | 1 - 2 | 163 | 61 | |
U88908.1 | - | 1 - 2 | 206 | 61 |