Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grn
Entrez Gene ID
14824
Gene Name
granulin
Gene Aliases
Pgrn, epithelin
Chromosome Location
Chr.11: 102430322 - 102436809 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008175.4 NP_032201.2 6 - 7 825 62
GenBank mRNA AK013678.1 - 6 - 7 641 62
AK018744.1 - 6 - 7 666 62
AK147932.1 - 6 - 7 63
AK149570.1 - 6 - 7 644 62
AK150125.1 - 6 - 7 640 62
AK150205.1 - 6 - 7 641 62
AK150338.1 - 6 - 7 630 62
AK150420.1 - 6 - 7 629 62
AK150717.1 - 6 - 7 641 62
AK150746.1 - 6 - 7 642 62
AK150881.1 - 6 - 7 642 62
AK151007.1 - 6 - 7 641 62
AK151162.1 - 6 - 7 642 62
AK151343.1 - 6 - 7 641 62
AK151713.1 - 6 - 7 641 62
AK151756.1 - 6 - 7 641 62
AK152048.1 - 6 - 7 641 62
AK152087.1 - 6 - 7 641 62
AK152118.1 - 6 - 7 630 62
AK152213.1 - 6 - 7 630 62
AK152705.1 - 6 - 7 641 62
AK152765.1 - 6 - 7 641 62
AK152944.1 - 6 - 7 630 62
AK153183.1 - 6 - 7 630 62
AK153269.1 - 6 - 7 641 62
AK153289.1 - 6 - 7 641 62
AK153418.1 - 6 - 7 641 62
AK153470.1 - 6 - 7 641 62
AK154492.1 - 6 - 7 641 62
AK159397.1 - 6 - 7 630 62
AK159428.1 - 6 - 7 630 62
AK159557.1 - 6 - 7 641 62
AK159809.1 - 6 - 7 641 62
AK160033.1 - 6 - 7 641 62
AK160074.1 - 6 - 7 630 62
AK211032.1 - 1 - 2 93 62
AK219696.1 - 1 - 2 104 62
BC037047.1 - 5 - 6 622 62
BC049943.1 - 6 - 7 627 62
BC129849.1 - 6 - 7 625 62
BC129850.1 - 6 - 7 625 62
X62321.1 - 5 - 6 606 62

View Full Product Details