The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277265.1 | NP_001264194.1 | 2 - 3 | 1078 | 63 |
NM_009325.4 | NP_033351.1 | 3 - 4 | 1232 | 63 | |
GenBank mRNA | AB115920.1 | - | 1 - 2 | 776 | 63 |
AK158477.1 | - | 2 - 3 | 1051 | 63 | |
BC089611.1 | - | 3 - 4 | 1205 | 63 | |
BC138885.1 | - | 2 - 3 | 1013 | 63 | |
BC138886.1 | - | 2 - 3 | 1013 | 63 | |
D10849.1 | - | 2 - 3 | 957 | 63 |