Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcna1
Entrez Gene ID
16485
Gene Name
potassium voltage-gated channel, shaker-related subfamily, member 1
Gene Aliases
AI840627, Kca1-1, Kv1.1, MBK1, Mk-1, Shak, mceph
Chromosome Location
Chr.6: 126636463 - 126645801 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010595.3 NP_034725.3 1 - 1 1038 122
GenBank mRNA AK039559.1 - 1 - 1 34 122
Y00305.1 - 1 - 1 50 122

View Full Product Details