Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pck1
Entrez Gene ID
18534
Gene Name
phosphoenolpyruvate carboxykinase 1, cytosolic
Gene Aliases
AI265463, PEPCK, Pck-1
Chromosome Location
Chr.2: 173153073 - 173159254 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011044.2 NP_035174.1 4 - 5 750 98
GenBank mRNA AK028046.1 - 4 - 5 751 98
AK030327.1 - 4 - 5 750 98
AK133496.1 - 4 - 5 751 98
AK149525.1 - 4 - 5 751 98
BC028342.1 - 4 - 5 752 98
BC037629.1 - 4 - 5 745 98

View Full Product Details