Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgef7
Entrez Gene ID
54126
Gene Name
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location
Chr.8: 11728105 - 11835219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113517.1 NP_001106989.1 15 - 16 1867 72
NM_001113518.1 NP_001106990.1 14 - 15 1656 72
NM_017402.4 NP_059098.2 14 - 15 1656 72
GenBank mRNA AF247654.1 - 14 - 15 1552 72
AF343877.1 - 14 - 15 1536 72
AK052545.1 - 14 - 15 1627 72
AK129064.1 - 15 - 16 1803 72
AK139156.1 - 15 - 16 1796 72
AY220301.1 - 14 - 15 1621 72
BC044838.1 - 14 - 15 1641 72
U96634.1 - 12 - 13 1346 72

View Full Product Details