Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
59042
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
1110005D17Rik, Cope1
Chromosome Location
Chr.8: 70302785 - 70312990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021538.1 NP_067513.1 9 - 10 899 61
GenBank mRNA AB039837.1 - 9 - 10 899 61
AK003458.1 - 9 - 10 916 61
AK140753.1 - 7 - 8 2620 61
AK217937.1 - 2 - 3 137 61
BC009170.1 - 9 - 10 907 61
BC057328.1 - 7 - 8 2585 61
BC083336.1 - 9 - 10 881 61

View Full Product Details