The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164248.1 | NP_001157720.1 | 6 - 7 | 717 | 99 |
NM_001164249.1 | NP_001157721.1 | 6 - 7 | 717 | 99 | |
NM_001164252.1 | NP_001157724.1 | 5 - 6 | 690 | 99 | |
NM_001164254.1 | NP_001157726.1 | 5 - 6 | 690 | 99 | |
NM_001164255.1 | NP_001157727.1 | 6 - 7 | 717 | 99 | |
NM_001164256.1 | NP_001157728.1 | 5 - 6 | 690 | 99 | |
NM_024427.4 | NP_077745.2 | 6 - 7 | 717 | 99 | |
GenBank mRNA | AK002271.1 | - | 6 - 7 | 718 | 99 |
AK003175.1 | - | 6 - 7 | 718 | 99 | |
AK032942.1 | - | 6 - 7 | 766 | 99 | |
AK168640.1 | - | 6 - 7 | 714 | 99 | |
AK168783.1 | - | 6 - 7 | 714 | 99 | |
AK169188.1 | - | 6 - 7 | 718 | 99 | |
BC026720.1 | - | 6 - 7 | 691 | 99 | |
BC132035.1 | - | 6 - 7 | 676 | 99 | |
BC132037.1 | - | 6 - 7 | 676 | 99 | |
BC145307.1 | - | 6 - 7 | 676 | 99 | |
M22479.1 | - | 6 - 7 | 690 | 99 | |
X64831.1 | - | 6 - 7 | 672 | 99 |