The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164226.1 | NP_001157698.1 | 10 - 11 | 1520 | 62 |
NM_001164227.1 | NP_001157699.1 | 10 - 11 | 1520 | 62 | |
NM_001164229.1 | NP_001157701.1 | 11 - 12 | 1594 | 62 | |
NM_001164230.1 | NP_001157702.1 | 10 - 11 | 1517 | 62 | |
NM_010938.4 | NP_035068.3 | 10 - 11 | 1517 | 62 | |
GenBank mRNA | AF098077.1 | - | 10 - 11 | 1452 | 62 |
AK014494.1 | - | 10 - 11 | 1481 | 62 | |
AK029034.1 | - | 10 - 11 | 1498 | 62 | |
AK031103.1 | - | 5 - 6 | 721 | 62 | |
AK037697.1 | - | 10 - 11 | 1478 | 62 | |
AK082580.1 | - | 10 - 11 | 1518 | 62 | |
AK134483.1 | - | 10 - 11 | 1434 | 62 | |
AK135673.1 | - | 11 - 12 | 1597 | 62 | |
BC005410.1 | - | 9 - 10 | 1338 | 62 |