The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020626.2 | NP_065651.1 | 3 - 4 | 287 | 72 |
GenBank mRNA | AF178085.1 | - | 3 - 4 | 287 | 72 |
AK002337.1 | - | 3 - 4 | 233 | 72 | |
AK148534.1 | - | 3 - 4 | 284 | 72 | |
BC049912.1 | - | 3 - 4 | 209 | 72 |