The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053266.1 | NP_444496.1 | 3 - 4 | 459 | 71 |
GenBank mRNA | AK081801.1 | - | 3 - 4 | 468 | 71 |
AK082488.1 | - | - | 628 | 71 | |
AY014963.1 | - | 3 - 4 | 459 | 71 | |
AY116023.1 | - | 3 - 4 | 470 | 71 | |
BC132157.1 | - | 3 - 4 | 367 | 71 | |
BC132159.1 | - | 3 - 4 | 367 | 71 |