Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd4
Entrez Gene ID
96935
Gene Name
sushi domain containing 4
Gene Aliases
AI848994, E430021N18Rik, N28096
Chromosome Location
Chr.1: 182763865 - 182896596 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC021842.1 - 4 - 5 873 120

View Full Product Details