The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163024.1 | NP_001156496.1 | 35 - 36 | 5531 | 71 |
NM_001163025.1 | NP_001156497.1 | 33 - 34 | 5513 | 71 | |
NM_153395.2 | NP_700444.2 | 33 - 34 | 5510 | 71 | |
GenBank mRNA | AB014059.1 | - | 33 - 34 | 5032 | 71 |
AK011826.1 | - | 12 - 13 | 2300 | 71 | |
AK012182.2 | - | 13 - 14 | 2449 | 71 | |
AK122425.1 | - | 35 - 36 | 5531 | 71 | |
AK162859.1 | - | 10 - 11 | 2004 | 71 | |
BC048406.1 | - | 22 - 23 | 3307 | 71 | |
BC150943.1 | - | 33 - 34 | 5034 | 71 | |
CN702930.1 | - | 3 - 4 | 366 | 71 |