Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ago3
Entrez Gene ID
214150
Gene Name
argonaute RISC catalytic subunit 3
Gene Aliases
AW048688, C130014L07Rik, Eif2c3
Chromosome Location
Chr.4: 126326990 - 126434830 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153402.2 NP_700451.2 1 - 2 277 101
GenBank mRNA AB081473.1 - 1 - 2 129 101
AK035042.1 - 1 - 2 243 101
AK042049.1 - 1 - 2 199 101
AK083652.1 - 1 - 2 244 101
AK154436.1 - 1 - 2 278 101
AK171118.1 - 1 - 2 278 101
AK172226.1 - 1 - 2 199 101
BC137964.1 - 1 - 2 124 101
BC145061.1 - 1 - 2 124 101
BC145063.1 - 1 - 2 124 101
BY235425.1 - 1 - 2 291 101

View Full Product Details