The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039194.3 | NP_001034283.1 | 5 - 6 | 618 | 89 |
NM_001284300.1 | NP_001271229.1 | 5 - 6 | 618 | 89 | |
NM_153779.2 | NP_722474.2 | 5 - 6 | 611 | 89 | |
NM_178058.4 | NP_835159.1 | 5 - 6 | 618 | 89 | |
GenBank mRNA | AK017403.1 | - | 5 - 6 | 616 | 89 |
AK085656.1 | - | 5 - 6 | 570 | 89 | |
AK147741.1 | - | 5 - 6 | 595 | 89 | |
AK155240.1 | - | 5 - 6 | 558 | 89 | |
BC038129.1 | - | 5 - 6 | 611 | 89 |