The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031392.2 | NP_113569.1 | 1 - 2 | 178 | 78 |
GenBank mRNA | AB041854.1 | - | 1 - 2 | 247 | 78 |
AF348591.1 | - | 1 - 2 | 174 | 78 | |
AK037181.1 | - | 1 - 2 | 155 | 78 | |
AK046119.1 | - | 1 - 2 | 145 | 78 | |
AK052235.1 | - | 1 - 2 | 117 | 78 | |
AK146617.1 | - | 1 - 2 | 135 | 78 | |
AK163752.1 | - | 1 - 2 | 111 | 78 | |
AK165935.1 | - | 1 - 2 | 142 | 78 | |
AK166265.1 | - | 1 - 2 | 155 | 78 | |
AK169357.1 | - | 1 - 2 | 179 | 78 | |
BC050894.1 | - | 1 - 2 | 178 | 78 | |
BC054367.1 | - | 1 - 2 | 106 | 78 |