Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr6
Entrez Gene ID
83669
Gene Name
WD repeat domain 6
Gene Aliases
mWDR6
Chromosome Location
Chr.9: 108572313 - 108578670 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031392.2 NP_113569.1 3 - 4 2756 65
GenBank mRNA AB041854.1 - 3 - 4 2822 65
AF348591.1 - 3 - 4 2752 65
AK037181.1 - 3 - 4 2733 65
AK046119.1 - 3 - 4 2724 65
AK052235.1 - 3 - 4 2697 65
AK146617.1 - 3 - 4 2713 65
AK163752.1 - 3 - 4 2689 65
AK165935.1 - 3 - 4 2720 65
AK166265.1 - 3 - 4 2734 65
AK169357.1 - 3 - 4 2757 65
BC035951.1 - 2 - 3 735 65
BC050894.1 - 3 - 4 2756 65
BC054367.1 - 3 - 4 2684 65

View Full Product Details