Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g2e
Entrez Gene ID
26970
Gene Name
phospholipase A2, group IIE
Gene Aliases
-
Chromosome Location
Chr.4: 138877911 - 138882817 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012044.2 NP_036174.1 2 - 3 205 102
GenBank mRNA AF112984.1 - 2 - 3 205 102
AF166098.1 - 2 - 3 201 102
AK148187.1 - - 221 102
BC027524.1 - 1 - 2 94 102

View Full Product Details