The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008065.2 | NP_032091.2 | 3 - 4 | 668 | 94 |
GenBank mRNA | AK139916.1 | - | 3 - 4 | 348 | 94 |
AK145446.1 | - | 3 - 4 | 348 | 94 | |
AK145838.1 | - | 3 - 4 | 413 | 94 | |
AK148087.1 | - | 3 - 4 | 439 | 94 | |
AK150455.1 | - | 3 - 4 | 348 | 94 | |
BC013562.1 | - | 1 - 2 | 63 | 94 | |
BC052448.1 | - | 3 - 4 | 668 | 94 | |
M74515.1 | - | 3 - 4 | 675 | 94 |