Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gabpa
Entrez Gene ID
14390
Gene Name
GA repeat binding protein, alpha
Gene Aliases
GABPalpha
Chromosome Location
Chr.16: 84834925 - 84863783 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008065.2 NP_032091.2 7 - 8 1253 72
GenBank mRNA AK139916.1 - 7 - 8 933 72
AK145446.1 - 7 - 8 933 72
AK145838.1 - 7 - 8 998 72
AK148087.1 - 7 - 8 1024 72
AK150455.1 - 7 - 8 933 72
BC013562.1 - 5 - 6 648 72
BC052448.1 - 7 - 8 1253 72
FN545817.1 - 2 - 3 90 72
M74515.1 - 7 - 8 1260 72

View Full Product Details