The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079875.2 | NP_001073344.1 | 4 - 5 | 1370 | 100 |
NM_009241.3 | NP_033267.2 | 3 - 4 | 1364 | 100 | |
GenBank mRNA | AK005638.1 | - | 4 - 5 | 1357 | 100 |
AY228460.1 | - | 2 - 3 | 1045 | 100 | |
AY920282.1 | - | 3 - 4 | 1364 | 100 | |
AY920283.1 | - | 4 - 5 | 1343 | 100 | |
U33958.1 | - | 3 - 4 | 1354 | 100 |