Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mefv
Entrez Gene ID
54483
Gene Name
Mediterranean fever
Gene Aliases
FMF, TRIM20, pyrin
Chromosome Location
Chr.16: 3707215 - 3718210 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161790.1 NP_001155262.1 3 - 4 1622 84
NM_001161791.1 NP_001155263.1 3 - 4 1532 84
NM_019453.2 NP_062326.2 3 - 4 1532 84
GenBank mRNA AF143409.1 - 3 - 4 1505 84
BC108993.1 - 3 - 4 1493 84
BC108994.1 - 3 - 4 1583 84

View Full Product Details