The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021374.5 | NP_067349.2 | 1 - 2 | 227 | 101 |
GenBank mRNA | AF191552.1 | - | 1 - 2 | 117 | 101 |
AK013773.1 | - | 1 - 2 | 56 | 101 | |
AK047734.1 | - | 1 - 2 | 68 | 101 | |
AK158864.1 | - | 1 - 2 | 68 | 101 | |
BC116925.1 | - | 1 - 2 | 117 | 101 | |
BC116929.1 | - | 1 - 2 | 117 | 101 | |
BY127242.1 | - | 1 - 2 | 227 | 101 |